Assay Details
Gene Symbol: MUSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130715.2 | XP_014986201.1 | 105 | |||
XM_015130716.2 | XP_014986202.1 | 105 | ||||
GenBank mRNA | JU321378.1 | - | 105 | |||
JV506312.1 | - | 105 | ||||
JV529840.1 | - | 208 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
