Assay Details
Gene Symbol: BEND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098619.4 | XP_001098619.2 | 588 | 52 | ||
XM_015138295.2 | XP_014993781.1 | 587 | 52 | |||
XM_028848421.1 | XP_028704254.1 | 589 | 52 | |||
XM_028848422.1 | XP_028704255.1 | 589 | 52 | |||
GenBank mRNA | JV060733.1 | - | 35 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
