Assay Details
| Gene Symbol: TAFA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261362.1 | NP_001248291.1 | 74 | 72 | ||
| XM_015130404.2 | XP_014985890.1 | 557 | 72 | |||
| GenBank mRNA | JU320855.1 | - | 74 | 72 | ||
| JV600935.1 | - | 164 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261362.1, XM_015130404.2 transcripts.
Set Membership: |
|

TAFA1
Genomic Map