Assay Details
Gene Symbol: KCNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102114.4 | XP_001102114.2 | 1960 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
