Assay Details
| Gene Symbol: HTR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198700.2 | NP_001185629.1 | 1642 | 124 | ||
| GenBank mRNA | JV466910.1 | - | 749 | 124 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTR1A| Gene Symbol: HTR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198700.2 | NP_001185629.1 | 1642 | 124 | ||
| GenBank mRNA | JV466910.1 | - | 749 | 124 | ||
Set Membership: |
|