Assay Details
Gene Symbol: ADORA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095531.4 | XP_001095531.1 | 1138 | 98 | ||
XM_015149913.2 | XP_015005399.1 | 1255 | 98 | |||
XM_015149914.2 | XP_015005400.1 | 798 | 98 | |||
XM_015149915.2 | XP_015005401.1 | 927 | 98 | |||
XM_028828632.1 | XP_028684465.1 | 1337 | 98 | |||
GenBank mRNA | JU480495.1 | - | 790 | 98 | ||
JV707160.1 | - | 748 | 98 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
