Assay Details
| Gene Symbol: POMC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082745.4 | XP_001082745.1 | 553 | 140 | ||
| XM_001082874.4 | XP_001082874.1 | 533 | 140 | |||
| XM_001082999.4 | XP_001082999.1 | 548 | 140 | |||
| XM_015111836.2 | XP_014967322.1 | 583 | 140 | |||
More Information
Set Membership: |
|

POMC
Genomic Map