Assay Details
| Gene Symbol: PRRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124583.2 | XP_014980069.1 | 1325 | 105 | ||
| XM_015124584.2 | XP_014980070.1 | 724 | 105 | |||
More Information
Set Membership: |
|

PRRG2
Genomic Map