Assay Details
| Gene Symbol: PTGER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033033.1 | NP_001028205.1 | 944 | 141 | ||
| XM_015123080.2 | XP_014978566.2 | 3277 | 141 | |||
| GenBank mRNA | AJ884597.1 | - | 944 | 141 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PTGER1
Genomic Map