Assay Details
Gene Symbol: OR1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193987.2 | NP_001180916.2 | 831 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
