Assay Details
| Gene Symbol: TCEAL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127869.2 | XP_014983355.1 | 68 | 82 | ||
| XM_028841673.1 | XP_028697506.1 | 66 | 82 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015127869.2, XM_028841673.1 transcripts.
Set Membership: |
|

TCEAL9
Genomic Map