Assay Details
| Gene Symbol: LOC703511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128572.2 | XP_014984058.1 | 1641 | 63 | ||
| GenBank mRNA | JU326712.1 | - | 314 | 63 | ||
| JV662774.1 | - | 662 | 63 | |||
| Gene Symbol: LOC114675039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842397.1 | XP_028698230.1 | 1062 | 63 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC703511
Genomic Map