Assay Details
Gene Symbol: HMGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134782.1 | XP_014990268.1 | 1030 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015134782.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
