Assay Details
| Gene Symbol: CALHM6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111520.4 | XP_001111520.1 | 778 | 145 | ||
| GenBank mRNA | JX502179.1 | - | 506 | 145 | ||
| JX502180.1 | - | 506 | 145 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CALHM6
Genomic Map