Assay Details
| Gene Symbol: XCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114298.3 | XP_001114298.2 | 398 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001114298.3 transcripts.
Set Membership: |
|

XCR1
Genomic Map