Assay Details
| Gene Symbol: CPTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257474.2 | NP_001244403.1 | 453 | 96 | ||
| GenBank mRNA | JU332272.1 | - | 453 | 96 | ||
| JU472357.1 | - | 466 | 96 | |||
| JV045710.1 | - | 175 | 96 | |||
| JV637005.1 | - | 537 | 96 | |||
More Information
Set Membership: |
|

CPTP
Genomic Map