Assay Details
Gene Symbol: SSTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085574.4 | XP_001085574.3 | 976 | 55 | ||
GenBank mRNA | JU336077.1 | - | 99 | 55 | ||
JV626838.1 | - | 259 | 55 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001085574.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
