Assay Details
Gene Symbol: CEBPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194337.2 | NP_001181266.1 | 1039 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
