Assay Details
| Gene Symbol: JUNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194461.1 | NP_001181390.1 | 79 | 119 | ||
| GenBank mRNA | JU321779.1 | - | 60 | 119 | ||
| JU473310.1 | - | 60 | 119 | |||
| JV601257.1 | - | 65 | 119 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194461.1 transcripts.
Set Membership: |
|

JUNB
Genomic Map