Assay Details
Gene Symbol: KCNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258108.1 | NP_001245037.1 | 2905 | 120 | ||
XM_015146956.2 | XP_015002442.1 | 3042 | 120 | |||
XM_015146960.2 | XP_015002446.1 | 3042 | 120 | |||
GenBank mRNA | JU475508.1 | - | 2909 | 120 | ||
JU475509.1 | - | 2905 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
