Assay Details
Gene Symbol: KCNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102294.4 | XP_001102294.2 | 2708 | 150 | ||
GenBank mRNA | JV714264.1 | - | 640 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
