Assay Details
Gene Symbol: KCNA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001101937.4 | XP_001101937.1 | 790 | 136 | ||
XM_015150880.2 | XP_015006366.1 | 790 | 136 | |||
XM_028829177.1 | XP_028685010.1 | 790 | 136 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001101937.4, XM_015150880.2, XM_028829177.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
