Assay Details
| Gene Symbol: NDUFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139246.2 | XP_014994732.1 | 467 | 148 | ||
| XM_028848780.1 | XP_028704613.1 | 463 | 148 | |||
| GenBank mRNA | JU323474.1 | - | 206 | 148 | ||
| JU473659.1 | - | 333 | 148 | |||
| JV047150.1 | - | 382 | 148 | |||
| JV684952.1 | - | 128 | 148 | |||
More Information
Set Membership: |
|

NDUFC1
Genomic Map