Assay Details
| Gene Symbol: TNFSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101190.4 | XP_001101190.1 | 932 | 146 | ||
| XM_015123968.2 | XP_014979454.1 | 1185 | 146 | |||
| XM_015123982.2 | XP_014979468.1 | 709 | 146 | |||
| XM_015123991.2 | XP_014979477.1 | 869 | 146 | |||
| XM_015123997.2 | XP_014979483.1 | 643 | 146 | |||
| XM_015124003.2 | XP_014979489.1 | 1918 | 146 | |||
| XM_028850506.1 | XP_028706339.1 | 962 | 146 | |||
| XM_028850527.1 | XP_028706360.1 | 1673 | 146 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TNFSF4
Genomic Map