Assay Details
| Gene Symbol: WNT9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114592.2 | XP_014970078.1 | 4619 | 105 | ||
| XM_015114594.2 | XP_014970080.1 | 698 | 105 | |||
| GenBank mRNA | JU331891.1 | - | 42 | 105 | ||
More Information
Set Membership: |
|

WNT9A
Genomic Map