Assay Details
Gene Symbol: FZD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100283.4 | XP_001100283.2 | 3201 | 102 | ||
GenBank mRNA | JV724567.1 | - | 2273 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
