Assay Details
Gene Symbol: FZD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111034.4 | XP_001111034.3 | 1946 | 135 | ||
GenBank mRNA | JV630281.1 | - | 1574 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
