Assay Details
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267004.1 | NP_001253933.1 | 406 | 122 | ||
GenBank mRNA | JU336904.1 | - | 409 | 122 | ||
JU477647.1 | - | 406 | 122 | |||
JV043905.1 | - | 363 | 122 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
