Assay Details
Gene Symbol: TAAR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098401.1 | NP_001091871.1 | 950 | 117 | ||
GenBank mRNA | EF549701.1 | - | 950 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
