Assay Details
Gene Symbol: THAP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115225.2 | XP_014970711.2 | 3031 | 170 | ||
XM_028833832.1 | XP_028689665.1 | 2908 | 170 | |||
GenBank mRNA | JU327273.1 | - | 2724 | 170 | ||
JU476423.1 | - | 2741 | 170 | |||
JV587468.1 | - | 1350 | 170 | |||
JV636273.1 | - | 2737 | 170 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
