Assay Details
| Gene Symbol: KCNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265653.1 | NP_001252582.1 | 721 | 143 | ||
| GenBank mRNA | JV544027.1 | - | 499 | 143 | ||
| JV565113.1 | - | 721 | 143 | |||
| JV615865.1 | - | 728 | 143 | |||
More Information
Set Membership: |
|

KCNB1
Genomic Map