Assay Details
Gene Symbol: MSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082936.4 | XP_001082936.2 | 841 | 61 | ||
GenBank mRNA | JV075610.1 | - | 284 | 61 | ||
JV635859.1 | - | 669 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
