Assay Details
| Gene Symbol: TAF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193945.1 | NP_001180874.1 | 1958 | 112 | ||
| GenBank mRNA | JU320911.1 | - | 1954 | 112 | ||
| JU473582.1 | - | 1921 | 112 | |||
| JV043918.1 | - | 1876 | 112 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TAF7
Genomic Map