Assay Details
| Gene Symbol: TOPORS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266141.1 | NP_001253070.1 | 366 | 159 | ||
| GenBank mRNA | JU332447.1 | - | 364 | 159 | ||
| JU473914.1 | - | 366 | 159 | |||
More Information
Set Membership: |
|

TOPORS
Genomic Map