Assay Details
Gene Symbol: LOC114675571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842784.1 | XP_028698617.1 | 486 | 145 |
Gene Symbol: MC5R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193897.2 | NP_001180826.2 | 274 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
