Assay Details
Gene Symbol: CITED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137345.2 | XP_014992831.1 | 352 | 80 | ||
GenBank mRNA | JU324361.1 | - | 202 | 80 | ||
JV049572.1 | - | 239 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015137345.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
