Assay Details
| Gene Symbol: NKX3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266223.1 | NP_001253152.1 | 320 | 123 | ||
| GenBank mRNA | JV636177.1 | - | 320 | 123 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NKX3-1| Gene Symbol: NKX3-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266223.1 | NP_001253152.1 | 320 | 123 | ||
| GenBank mRNA | JV636177.1 | - | 320 | 123 | ||
Set Membership: |
|