Assay Details
| Gene Symbol: B3GNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261632.1 | NP_001248561.1 | 217 | 135 | ||
| GenBank mRNA | JU332575.1 | - | 136 | 135 | ||
| JU473507.1 | - | 227 | 135 | |||
| JV044708.1 | - | 217 | 135 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261632.1 transcripts.
Set Membership: |
|

B3GNT2
Genomic Map