Assay Details
| Gene Symbol: MLLT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260657.1 | NP_001247586.1 | 250 | 94 | ||
| GenBank mRNA | JU322629.1 | - | 487 | 94 | ||
| JU322630.1 | - | 487 | 94 | |||
| JU473456.1 | - | 250 | 94 | |||
| JV703561.1 | - | 704 | 94 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260657.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MLLT11
Genomic Map