Assay Details
Gene Symbol: DNAJB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277937.1 | NP_001264866.1 | 2090 | 90 | ||
XM_015142773.2 | XP_014998259.1 | 2174 | 90 | |||
XM_015142787.2 | XP_014998273.1 | 2295 | 90 | |||
XM_015142789.2 | XP_014998275.1 | 1996 | 90 | |||
XM_015142800.2 | XP_014998286.1 | 2171 | 90 | |||
XM_015142809.2 | XP_014998295.1 | 1843 | 90 | |||
XM_028843305.1 | XP_028699138.1 | 1966 | 90 | |||
GenBank mRNA | JU320708.1 | - | 2090 | 90 | ||
JU471858.1 | - | 2094 | 90 | |||
JV568056.1 | - | 2084 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
