Assay Details
Gene Symbol: LSM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266965.2 | NP_001253894.1 | 67 | 99 | ||
GenBank mRNA | JU319787.1 | - | 112 | 99 | ||
JV046059.1 | - | 69 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266965.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
