Assay Details
Gene Symbol: ZNF146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103038.4 | XP_001103038.2 | 226 | 184 | ||
XM_015123742.2 | XP_014979228.2 | 291 | 184 | |||
XM_015123743.2 | XP_014979229.2 | 365 | 184 | |||
XM_015123744.2 | XP_014979230.2 | 362 | 184 | |||
XM_015123745.2 | XP_014979231.2 | 288 | 184 | |||
XM_015123747.2 | XP_014979233.2 | 249 | 184 | |||
XM_028839608.1 | XP_028695441.1 | 362 | 184 | |||
XM_028839609.1 | XP_028695442.1 | 359 | 184 | |||
XM_028839610.1 | XP_028695443.1 | 208 | 184 | |||
XM_028839611.1 | XP_028695444.1 | 151 | 184 | |||
GenBank mRNA | JU320253.1 | - | 334 | 184 | ||
JU471202.1 | - | 365 | 184 | |||
JV043822.1 | - | 226 | 184 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001103038.4, XM_015123742.2, XM_015123743.2, XM_015123744.2, XM_015123745.2, XM_015123747.2, XM_028839608.1, XM_028839609.1, XM_028839610.1, XM_028839611.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
