Assay Details
Gene Symbol: LOC703511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128572.2 | XP_014984058.1 | 2166 | 142 | ||
GenBank mRNA | JU326712.1 | - | 839 | 142 | ||
JV662774.1 | - | 1187 | 142 |
Gene Symbol: LOC114675039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842397.1 | XP_028698230.1 | 1587 | 142 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
