Assay Details
Gene Symbol: TIMM10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109886.4 | XP_001109886.3 | 826 | 62 | ||
XM_028833758.1 | XP_028689591.1 | 826 | 62 | |||
GenBank mRNA | JV464563.1 | - | 77 | 62 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
