Assay Details
Gene Symbol: SERTAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261264.1 | NP_001248193.1 | 1041 | 130 | ||
GenBank mRNA | JU334223.1 | - | 1041 | 130 | ||
JU473695.1 | - | 1041 | 130 | |||
JV466390.1 | - | 669 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
