Assay Details
Gene Symbol: RAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122061.2 | XP_014977547.2 | 904 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
