Assay Details
Gene Symbol: UBQLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127555.2 | XP_014983041.1 | 303 | 69 | ||
GenBank mRNA | JU322065.1 | - | 100 | 69 | ||
JU475341.1 | - | 128 | 69 | |||
JV542819.1 | - | 128 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015127555.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
