Assay Details
| Gene Symbol: S100A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261799.1 | NP_001248728.1 | 275 | 83 | ||
| XM_028845294.1 | XP_028701127.1 | 264 | 83 | |||
| GenBank mRNA | JU320264.1 | - | 259 | 83 | ||
| JU473185.1 | - | 275 | 83 | |||
| JV658295.1 | - | 284 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

S100A6
Genomic Map