Assay Details
Gene Symbol: EPM2AIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090676.4 | XP_001090676.3 | 2985 | 96 | ||
XM_028843929.1 | XP_028699762.1 | 2985 | 96 | |||
GenBank mRNA | JU321008.1 | - | 2707 | 96 | ||
JU471327.1 | - | 2717 | 95 | |||
JV045019.1 | - | 2756 | 96 | |||
JV637256.1 | - | 2825 | 96 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
