Assay Details
| Gene Symbol: GLCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266557.1 | NP_001253486.1 | 127 | 139 | ||
| GenBank mRNA | JU333908.1 | - | 127 | 139 | ||
| JU475095.1 | - | 127 | 139 | |||
| JV647834.1 | - | 130 | 139 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266557.1 transcripts.
Set Membership: |
|

GLCE
Genomic Map